3-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}acrylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-[4-(2-Chloro-6-fluorobenzyloxy)phenyl]acrylic acid
043542-500MG
043542-1G
043542-5G
101861-354EA
375.38
USD
101861-354
101861-356
101861-358
3-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}acrylic acid ≥95%
3-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}acrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₂ClFO₃ Melting Pt: 177…179 °C |
MDL Number:
MFCD02091011 CAS Number: 400825-72-1 |
Specification Test Results
Melting point | 177-179°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...