3-{4-[(2,4-Dichlorobenzyl)oxy]phenyl}acrylic acid ≥95%

Supplier: Matrix Scientific

043541-500MG 043541-1G 043541-5G
101861-348EA 278.46 USD
101861-348 101861-350 101861-352
3-{4-[(2,4-Dichlorobenzyl)oxy]phenyl}acrylic acid ≥95%
3-{4-[(2,4-Dichlorobenzyl)oxy]phenyl}acrylic acid

Hazard: Irritant

Formula: C₁₆H₁₂Cl₂O₃
Melting Pt: 234…236 °C
MDL Number: MFCD03001248
CAS Number: 385383-41-5

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR