3-{4-[(3-Chlorobenzyl)oxy]phenyl}acrylic acid ≥95%

Supplier: Matrix Scientific

043539-500MG 043539-1G 043539-5G
101861-336EA 278.46 USD
101861-336 101861-338 101861-340
3-{4-[(3-Chlorobenzyl)oxy]phenyl}acrylic acid ≥95%
3-{4-[(3-Chlorobenzyl)oxy]phenyl}acrylic acid

Hazard: Irritant

Formula: C₁₆H₁₃ClO₃
Melting Pt: 210…212 °C
MDL Number: MFCD03001246
CAS Number: 385383-37-9

Order Now

Specification Test Results

Melting point 210-212°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR