3-[4-(Benzyloxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Benzyloxy)cinnamic acid
043538-500MG
043538-1G
043538-5G
101861-330EA
278.46
USD
101861-330
101861-332
101861-334
3-[4-(Benzyloxy)phenyl]acrylic acid ≥95%
3-[4-(Benzyloxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₃ MW: 254.29 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD00021778 CAS Number: 6272-45-3 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...