2-Piperidinecarboxamide ≥97%
Supplier: Matrix Scientific
043536-1G
043536-5G
101861-326EA
21.54
USD
101861-326
101861-328
2-Piperidinecarboxamide ≥97%
2-Piperidinecarboxamide
Hazard: Irritant
Formula:
C₆H₁₂N₂O Melting Pt: 138…140 °C |
MDL Number:
MFCD05663784 CAS Number: 19889-77-1 |
Specification Test Results
Melting point | 138-140°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...