4-(2,4-Dimethylphenoxy)benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
043534-500MG
043534-1G
043534-5G
101861-316EA
278.46
USD
101861-316
101861-318
101861-320
4-(2,4-Dimethylphenoxy)benzenecarbonitrile ≥95%
4-(2,4-Dimethylphenoxy)benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₅H₁₃NO Melting Pt: 62…64 °C |
MDL Number:
MFCD01815096 CAS Number: 449778-70-5 |
Specification Test Results
Melting point | 62-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...