3-[2-(4-Bromo-2-fluorophenoxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
043533-500MG
043533-1G
043533-5G
101861-310EA
556.92
USD
101861-310
101861-312
101861-314
3-[2-(4-Bromo-2-fluorophenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Bromo-2-fluorophenoxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₅H₁₀BrFO₃ Melting Pt: 219…221 °C |
MDL Number:
MFCD01814603 CAS Number: 449778-69-2 |
Specification Test Results
Melting point | 219-221°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...