2-(1-Phenylethylidene)-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

043532-500MG 043532-1G 043532-5G
101861-304EA 181.54 USD
101861-304 101861-306 101861-308
2-(1-Phenylethylidene)-1-hydrazinecarboxamide ≥95%
2-(1-Phenylethylidene)-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₉H₁₁N₃O
MW: 177.21 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD00137159
CAS Number: 2492-30-0

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR