4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(4-Chloro-3-fluorophenoxy)benzonitrile
043531-500MG
043531-1G
043531-5G
101861-298EA
375.38
USD
101861-298
101861-300
101861-302
4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile ≥95%
4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₃H₇ClFNO MW: 247.66 g/mol Melting Pt: 75…77 °C |
MDL Number:
MFCD01815095 CAS Number: 449778-68-1 |
Specification Test Results
Melting point | 75-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...