3-[2-(3,4-Difluorophenoxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

043528-500MG 043528-1G 043528-5G
101861-280EA 747.69 USD
101861-280 101861-282 101861-284
3-[2-(3,4-Difluorophenoxy)phenyl]acrylic acid ≥95%
3-[2-(3,4-Difluorophenoxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₅H₁₀F₂O₃
Melting Pt: 198…200 °C
MDL Number: MFCD02091009
CAS Number: 338393-76-3

Order Now

Specification Test Results

Melting point 198-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR