3-[2-(3,5-Dimethylphenoxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

043527-500MG 043527-1G 043527-5G
101861-274EA 556.92 USD
101861-274 101861-276 101861-278
3-[2-(3,5-Dimethylphenoxy)phenyl]acrylic acid ≥95%
3-[2-(3,5-Dimethylphenoxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₇H₁₆O₃
Melting Pt: 201…203 °C
MDL Number: MFCD02091008
CAS Number: 338393-75-2

Order Now

Specification Test Results

Melting point 201-203°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR