2-[4-(Benzyloxy)phenoxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043526-500MG 043526-1G 043526-5G
101861-268EA 250.77 USD
101861-268 101861-270 101861-272
2-[4-(Benzyloxy)phenoxy]benzenecarbaldehyde ≥95%
2-[4-(Benzyloxy)phenoxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₂₀H₁₆O₃
Melting Pt: 64…66 °C
MDL Number: MFCD01568899
CAS Number: 338393-68-3

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR