3-[2-(4-Fluorophenoxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

043524-500MG 043524-1G 043524-5G
101861-256EA 556.92 USD
101861-256 101861-258 101861-260
3-[2-(4-Fluorophenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Fluorophenoxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₅H₁₁FO₃
Melting Pt: 205…207 °C
MDL Number: MFCD02091006
CAS Number: 338393-64-9

Order Now

Specification Test Results

Melting point 205-207°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR