2-(4-Chloro-3-fluorophenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

043523-500MG 043523-1G
101861-252EA 1109.23 USD
101861-252 101861-254
2-(4-Chloro-3-fluorophenoxy)benzaldehyde ≥95%
2-(4-Chloro-3-fluorophenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClFO₂
Melting Pt: 66-68°C
MDL Number: MFCD01568894
CAS Number: 338393-57-0

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR