Methyl-3-[(2,4-dichlorobenzyl)oxy]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043522-500MG 043522-1G 043522-5G
101861-246EA 181.54 USD
101861-246 101861-248 101861-250
Methyl-3-[(2,4-dichlorobenzyl)oxy]benzenecarboxylate ≥95%
Methyl-3-[(2,4-dichlorobenzyl)oxy]benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₃
Melting Pt: 116…118 °C
MDL Number: MFCD01568893
CAS Number: 58041-99-9

Order Now

Specification Test Results

Melting point 116-118°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR