4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043520-500MG 043520-1G 043520-5G
101861-234EA 181.54 USD
101861-234 101861-236 101861-238
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde ≥95%
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₈ClNO₂S
Melting Pt: 107…108 °C
MDL Number: MFCD01568788
CAS Number: 338393-48-9

Order Now

Specification Test Results

Melting point 107-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR