2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
043519-500MG
043519-1G
043519-5G
101861-228EA
181.54
USD
101861-228
101861-230
101861-232
2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde ≥95%
2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₁H₈ClNO₂S Melting Pt: 75…77 °C |
MDL Number:
MFCD01568787 CAS Number: 338393-47-8 |
Specification Test Results
Melting point | 75-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...