Methyl-2-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
043518-500MG
043518-1G
101861-224EA
375.38
USD
101861-224
101861-226
Methyl-2-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate ≥95%
Methyl-2-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₃S Melting Pt: 62…64 °C |
MDL Number:
MFCD01568783 CAS Number: 338393-43-4 |
Specification Test Results
Melting point | 62-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...