Methyl-3-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
043517-500MG
043517-1G
043517-5G
101861-218EA
181.54
USD
101861-218
101861-220
101861-222
Methyl-3-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate ≥95%
Methyl-3-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₃S Melting Pt: 97…99 °C |
MDL Number:
MFCD01568782 CAS Number: 338393-42-3 |
Specification Test Results
Melting point | 97-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...