2-Methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

043510-500MG 043510-1G 043510-5G
101861-186EA 375.38 USD
101861-186 101861-188 101861-190
2-Methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
2-Methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 222…224 °C
MDL Number: MFCD00974996
CAS Number: 27036-90-4

Order Now

Specification Test Results

Melting point 222-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR