Ethyl-2-(3-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyridinyl)acetate ≥95%

Supplier: Matrix Scientific

043509-500MG 043509-1G 043509-5G
101861-180EA 375.38 USD
101861-180 101861-182 101861-184
Ethyl-2-(3-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyridinyl)acetate ≥95%
Ethyl-2-(3-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyridinyl)acetate

Hazard: Irritant

Formula: C₁₇H₁₅F₃N₂O₃
MW: 352.32 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00974992
CAS Number: 338392-12-4

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR