Ethyl-2-{3-[(4-fluoroanilino)carbonyl]-2-pyridinyl}acetate ≥95%

Supplier: Matrix Scientific

043508-500MG 043508-1G
101861-176EA 375.38 USD
101861-176 101861-178
Ethyl-2-{3-[(4-fluoroanilino)carbonyl]-2-pyridinyl}acetate ≥95%
Ethyl-2-{3-[(4-fluoroanilino)carbonyl]-2-pyridinyl}acetate

Hazard: Irritant

Formula: C₁₆H₁₅FN₂O₃
MW: 302.31 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD00974991
CAS Number: 338392-11-3

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR