3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Hydroxyprop-1-yl)-3-(trifluoromethyl)-1H-pyrazole

043507-500MG 043507-1G 043507-5G
101861-170EA 269.23 USD
101861-170 101861-172 101861-174
3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol ≥95%
3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol

Hazard: Irritant

Formula: C₇H₉F₃N₂O
MW: 194.16 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD00794530
CAS Number: 169213-78-9

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR