1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone ≥95%

Supplier: Matrix Scientific

043506-500MG 043506-1G 043506-5G
101861-164EA 278.46 USD
101861-164 101861-166 101861-168
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone ≥95%
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone

Hazard: Irritant

Formula: C₁₁H₁₂BrNO
MW: 254.13 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD00793408
CAS Number: 338392-04-4

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR