N-(3-Bromophenyl)-3-chloro-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

043505-500MG 043505-1G 043505-5G
101861-158EA 181.54 USD
101861-158 101861-160 101861-162
N-(3-Bromophenyl)-3-chloro-2,2-dimethylpropanamide ≥95%
N-(3-Bromophenyl)-3-chloro-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₁H₁₃BrClNO
Melting Pt: 108…110 °C
MDL Number: MFCD00793407
CAS Number: 338392-03-3

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR