1-(3-Phenyl-5-isoxazolyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(3-Phenylisoxazol-5yl)ethanone
043504-500MG
043504-1G
043504-5G
101861-152EA
556.92
USD
101861-152
101861-154
101861-156
1-(3-Phenyl-5-isoxazolyl)-1-ethanone ≥95%
1-(3-Phenyl-5-isoxazolyl)-1-ethanone
Hazard: Irritant
Formula:
C₁₁H₉NO₂ MW: 187.2 g/mol Melting Pt: 107…108.5 °C |
MDL Number:
MFCD00100033 CAS Number: 2048-69-3 |
Specification Test Results
Melting point | 107-108.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...