4-(5-Ethoxy-1,3-oxazol-2-yl)phenyl methyl ether ≥95%

Supplier: Matrix Scientific

043502-500MG 043502-1G 043502-5G
101861-140EA 375.38 USD
101861-140 101861-142 101861-144
4-(5-Ethoxy-1,3-oxazol-2-yl)phenyl methyl ether ≥95%
4-(5-Ethoxy-1,3-oxazol-2-yl)phenyl methyl ether

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
Melting Pt: 47…49 °C
MDL Number: MFCD00793751
CAS Number: 477867-59-7

Order Now

Specification Test Results

Melting point 47-49°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR