Ethyl-2-[(4-nitrobenzoyl)amino]acetate ≥95%
Supplier: Matrix Scientific
043501-500MG
043501-1G
043501-5G
101861-134EA
181.54
USD
101861-134
101861-136
101861-138
Ethyl-2-[(4-nitrobenzoyl)amino]acetate ≥95%
Ethyl-2-[(4-nitrobenzoyl)amino]acetate
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₅ Melting Pt: 147…149 °C |
MDL Number:
MFCD00024715 CAS Number: 7512-77-8 |
Specification Test Results
Melting point | 147-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...