Ethyl-2-[(4-methoxybenzoyl)amino]acetate ≥95%
Supplier: Matrix Scientific
043500-500MG
043500-1G
043500-5G
101861-128EA
181.54
USD
101861-128
101861-130
101861-132
Ethyl-2-[(4-methoxybenzoyl)amino]acetate ≥95%
Ethyl-2-[(4-methoxybenzoyl)amino]acetate
Hazard: Irritant
Formula:
C₁₂H₁₅NO₄ Melting Pt: 100…102 °C |
MDL Number:
MFCD00793747 CAS Number: 51220-57-6 |
Specification Test Results
Melting point | 100-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...