Ethyl-2-[(2-chlorobenzoyl)amino]acetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-[(2-chlorophenyl)formamido]acetate
043499-500MG
043499-1G
043499-5G
101861-122EA
181.54
USD
101861-122
101861-124
101861-126
Ethyl-2-[(2-chlorobenzoyl)amino]acetate ≥95%
Ethyl-2-[(2-chlorobenzoyl)amino]acetate
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₃ MW: 241.68 g/mol Melting Pt: 75…77 °C |
MDL Number:
MFCD00793745 CAS Number: 66824-94-0 |
Specification Test Results
Melting point | 75-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...