Ethyl-2-[(2-furylcarbonyl)amino]acetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(furan-2-carboxamido)acetate
043498-500MG
043498-1G
043498-5G
101861-116EA
181.54
USD
101861-116
101861-118
101861-120
Ethyl-2-[(2-furylcarbonyl)amino]acetate ≥95%
Ethyl-2-[(2-furylcarbonyl)amino]acetate
Hazard: Irritant
Formula:
C₉H₁₁NO₄ MW: 197.19 g/mol Melting Pt: 80…82 °C |
MDL Number:
MFCD00753958 CAS Number: 66318-15-8 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...