Ethyl-2-[(2-fluorobenzoyl)amino]acetate ≥95%
Supplier: Matrix Scientific
043497-500MG
043497-1G
043497-5G
101861-110EA
181.54
USD
101861-110
101861-112
101861-114
Ethyl-2-[(2-fluorobenzoyl)amino]acetate ≥95%
Ethyl-2-[(2-fluorobenzoyl)amino]acetate
Hazard: Irritant
Formula:
C₁₁H₁₂FNO₃ Melting Pt: 40…42 °C |
MDL Number:
MFCD00793744 CAS Number: 304657-05-4 |
Specification Test Results
Melting point | 40-42°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...