Ethyl-2-[(2,6-dichlorobenzoyl)amino]acetate ≥95%
Supplier: Matrix Scientific
043496-500MG
043496-1G
043496-5G
101861-104EA
181.54
USD
101861-104
101861-106
101861-108
Ethyl-2-[(2,6-dichlorobenzoyl)amino]acetate ≥95%
Ethyl-2-[(2,6-dichlorobenzoyl)amino]acetate
Hazard: Irritant
Formula:
C₁₁H₁₁Cl₂NO₃ Melting Pt: 108…110 °C |
MDL Number:
MFCD00793743 CAS Number: 329931-03-5 |
Specification Test Results
Melting point | 108-110°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...