6-{[(4-Chlorophenyl)sulfanyl]methyl}-2-methyl-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
043492-500MG
043492-1G
043492-5G
101861-080EA
181.54
USD
101861-080
101861-082
101861-084
6-{[(4-Chlorophenyl)sulfanyl]methyl}-2-methyl-4-pyrimidinol ≥95%
6-{[(4-Chlorophenyl)sulfanyl]methyl}-2-methyl-4-pyrimidinol
Hazard: Irritant
Formula:
C₁₂H₁₁ClN₂OS Melting Pt: 168…170 °C |
MDL Number:
MFCD08056612 CAS Number: 886361-45-1 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...