5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

043491-500MG 043491-1G 043491-5G
101861-074EA 747.69 USD
101861-074 101861-076 101861-078
5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde ≥95%
5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₉H₁₇BrN₂OS₂
Melting Pt: 83…85.5 °C
MDL Number: MFCD01935941
CAS Number: 400823-20-3

Order Now

Specification Test Results

Melting point 83-85.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR