5-Chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
043490-500MG
043490-1G
043490-5G
101861-068EA
747.69
USD
101861-068
101861-070
101861-072
5-Chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde ≥95%
5-Chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₃ClN₂OS Melting Pt: 93…95.5 °C |
MDL Number:
MFCD00794830 CAS Number: 321526-27-6 |
Specification Test Results
Melting point | 93-95.5°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...