5-(Aminomethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione ≥95%

Supplier: Matrix Scientific

043489-500MG 043489-1G 043489-5G
101861-062EA 160 USD
101861-062 101861-064 101861-066
5-(Aminomethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione ≥95%
5-(Aminomethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione

Hazard: Irritant

Formula: C₇H₉NO₄
Melting Pt: 210.5…211.5 °C
MDL Number: MFCD01111609
CAS Number: 15568-88-4

Order Now

Specification Test Results

Melting point 210.5-211.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR