5-(Methoxymethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione ≥95%
Supplier: Matrix Scientific
043488-1G
043488-5G
043488-25G
101861-058EA
64.62
USD
101861-058
101861-060
102551-318
5-(Methoxymethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione ≥95%
5-(Methoxymethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione
Hazard: Irritant/Stored under Argon/Store in Freezer
Formula:
C₈H₁₀O₅ Melting Pt: 132…134 °C |
MDL Number:
MFCD00599240 CAS Number: 15568-85-1 |
Specification Test Results
Melting point | 132-134°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...