7-[1-(2,6-Dichlorophenoxy)ethyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
Supplier: Matrix Scientific
043486-500MG
043486-1G
043486-5G
101861-044EA
747.69
USD
101861-044
101861-046
101861-048
7-[1-(2,6-Dichlorophenoxy)ethyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-[1-(2,6-Dichlorophenoxy)ethyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Hazard: Irritant
Formula:
C₁₃H₁₁Cl₂N₅O MW: 324.17 g/mol Melting Pt: 255…257 °C |
MDL Number:
MFCD00243624 CAS Number: 477865-30-8 |
Specification Test Results
Melting point | 255-257°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...