7-(1-Phenoxyethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

043485-500MG 043485-1G 043485-5G
101861-038EA 376.92 USD
101861-038 101861-040 101861-042
7-(1-Phenoxyethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-(1-Phenoxyethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₃H₁₃N₅O
Melting Pt: 219…221 °C
MDL Number: MFCD00243606
CAS Number: 477865-07-9

Order Now

Specification Test Results

Melting point 219-221°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR