1-Methyl-3-phenyl-1H-pyrazol-5-ol ≥95%
Supplier: Matrix Scientific
043483-500MG
043483-1G
043483-5G
101861-028EA
181.54
USD
101861-028
101861-030
101861-032
1-Methyl-3-phenyl-1H-pyrazol-5-ol ≥95%
1-Methyl-3-phenyl-1H-pyrazol-5-ol
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O Melting Pt: 213…215 °C |
MDL Number:
MFCD03001245 CAS Number: 34347-81-4 |
Specification Test Results
Melting point | 213-215°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...