2-Oxo-6-phenyl-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
1,2-Dihydro-2-oxo-6-phenyl-4-(trifluoromethyl)pyridine-3-carbonitrile
043481-500MG
043481-1G
043481-5G
101861-022EA
181.54
USD
101861-022
101861-024
101861-026
2-Oxo-6-phenyl-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
2-Oxo-6-phenyl-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₃H₇F₃N₂O MW: 264.21 g/mol Melting Pt: 300…302 °C |
MDL Number:
MFCD00474273 CAS Number: 3335-44-2 |
Specification Test Results
Melting point | 300-302°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...