4-(2-sulfanyl-4,5-dihydro-1H-imidazol-1-yl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
043480-500MG
043480-1G
043480-5G
101861-016EA
375.38
USD
101861-016
101861-018
101861-020
4-(2-sulfanyl-4,5-dihydro-1H-imidazol-1-yl)benzenecarboxylic acid ≥95%
4-(2-sulfanyl-4,5-dihydro-1H-imidazol-1-yl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂S Melting Pt: 260…270 °C |
MDL Number:
MFCD00664192 CAS Number: 148720-11-0 |
Specification Test Results
Melting point | 260-270°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...