4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-5-methoxy-2-(pyridin-2-yl)pyrimidine, 4-Chloro-5-methoxy-2-(2-pyridyl)pyrimidine

043476-250MG 043476-1G 043476-5G
101860-992EA 455.38 USD
101860-992 101860-994 101860-996
4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine ≥95%
4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₈ClN₃O
MW: 221.644 g/mol
Melting Pt: 168…171 °C
MDL Number: MFCD00215412
CAS Number: 321432-82-0

Order Now

Specification Test Results

Melting point 168-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR