4-[(Phenylsulfanyl)methyl]aniline ≥95%

Supplier: Matrix Scientific

043469-500MG 043469-1G 043469-5G
101860-958EA 278.46 USD
101860-958 101860-960 101860-962
4-[(Phenylsulfanyl)methyl]aniline ≥95%
4-[(Phenylsulfanyl)methyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₃NS
Melting Pt: 73…75 °C
MDL Number: MFCD00739202
CAS Number: 13738-70-0

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR