4-{[(4-Chlorophenyl)sulfanyl]methyl}phenylamine ≥95%
Supplier: Matrix Scientific
043468-500MG
043468-1G
043468-5G
101860-952EA
556.92
USD
101860-952
101860-954
101860-956
4-{[(4-Chlorophenyl)sulfanyl]methyl}phenylamine ≥95%
4-{[(4-Chlorophenyl)sulfanyl]methyl}phenylamine
Hazard: Irritant
Formula:
C₁₃H₁₂ClNS Melting Pt: 98…100 °C |
MDL Number:
MFCD00172478 CAS Number: 6969-14-8 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...