1-(Dimethylamino)-4-(3-pyridinyloxy)-1-penten-3-one ≥95%
Supplier: Matrix Scientific
043467-500MG
043467-1G
101860-948EA
278.46
USD
101860-948
101860-950
1-(Dimethylamino)-4-(3-pyridinyloxy)-1-penten-3-one ≥95%
1-(Dimethylamino)-4-(3-pyridinyloxy)-1-penten-3-one
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ MW: 220.27 g/mol Melting Pt: 101…104 °C |
MDL Number:
MFCD08056611 CAS Number: 886361-43-9 |
Specification Test Results
Melting point | 101-104°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...