{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrol-2-yl}methanol ≥95%

Supplier: Matrix Scientific

043466-500MG 043466-1G 043466-5G
101860-942EA 921.54 USD
101860-942 101860-944 101860-946
{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrol-2-yl}methanol ≥95%
{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrol-2-yl}methanol

Hazard: Irritant

Formula: C₁₁H₈ClF₃N₂O
Melting Pt: 96…98 °C
MDL Number: MFCD00231916
CAS Number: 321430-86-8

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR