1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbohydrazide ≥95%

Supplier: Matrix Scientific

043464-500MG 043464-1G 043464-5G
101860-930EA 1109.23 USD
101860-930 101860-932 101860-934
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbohydrazide ≥95%
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₄ClF₃N₄O
MW: 322.72 g/mol
Melting Pt: 198…201 °C
MDL Number: MFCD00172276
CAS Number: 321430-54-0

Order Now

Specification Test Results

Melting point 198-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR