Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
043463-500MG
043463-1G
043463-5G
101860-924EA
747.69
USD
101860-924
101860-926
101860-928
Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₄ClF₃N₂O₂ MW: 322.72 g/mol Melting Pt: 52…54 °C |
MDL Number:
MFCD00172275 CAS Number: 321430-51-7 |
Specification Test Results
Melting point | 52-54°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...